Would you like to log in again?
Inlet Maintenance Kit for Agilent GCs, Restek
Supplier: RESTEK CORP
22181
10803-276EA 808.87 USD InStock
10803-276
Inlet Maintenance Kit for Agilent GCs, Restek
Tool Sets
For use with Agilent 5890, 6850, 6890, and 7890 GCs.
  • Kit includes the most common consumable GC supplies and tools
  • Parts list makes reordering easy
  • All parts meet or exceed performance of instrument manufacturer's parts
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty