Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
ALTEF® Gas Sampling Bags, Restek
Supplier: Restek
22959
10803-304PK 301.55 USD InStock
10803-304
ALTEF® Gas Sampling Bags, Restek
Bags Gas Bags
Sampling bags are a low-cost whole air sampling device for high level VOCs and permanent gases.
  • Excellent low-cost alternative to Tedlar® bags for collection of most VOCs
  • Very low VOC and sulfur background compared to Tedlar® bags
  • Not recommended for ketones, acetates, hydrogen sulfide, or permanent gases
  • Durable PVDF film is resistant to abrasion and chemicals, including most acids and organic compounds
  • Contain no additives, fillers, or pigments
Several EPA, NIOSH, and OSHA methods exist for bag sampling for a variety of applications: stationary sources emissions, workplace atmospheres, ambient, indoor air quality, and breath analysis. Choose the film type appropriate for the application. All Restek's bags feature a polypropylene combo valve with hose connection to fit ³/₁₆" Int. Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty