Would you like to log in again?
Ottawa Sand (Bulk Adsorbent), Restek
Supplier: Restek
26137
10803-396EA 315.65 USD InStock
10803-396
Ottawa Sand (Bulk Adsorbent), Restek
Spill Control Materials
Ottawa sand is listed in several U.S
  • Sample medium for matrix spikes and laboratory control blanks
  • Packaged in convenient 5 kg buckets
EPA methods as the specified medium for matrix spike and laboratory control blanks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty