Would you like to log in again?
MXT-1HT SIMDIST
Supplier: Avantor
Column^ MXT*-1HT Sim Dist (metal, Siltek* treated) Capillary Columns, 5m length, 0.53mm I.D, 0.10um df, Temperature Limits: -60 Degree to +430 Degree C, Material: Stainless Steel, Technique Crossbond*, Stationary Phase: Mxt*-1HT Sim Dist
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MXT-1HT SIMDIST 10803-428   Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty