Would you like to log in again?
CAPS SCREW SOFT GUARD SIL/PTFE RD SLIT P
Supplier: MicroSolv Technology Corp
Cap^ Cap screw top with single slit soft guard white silicone and red Polytetrafluoroethylene septa bonded in ribbed cap red for use with 9-425mm 12x32mm autosampler vials basik*, 100/pack
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS SCREW SOFT GUARD SIL/PTFE RD SLIT P 10803-690 9502S-E-30C-R Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty