Would you like to log in again?
NUT MALE FLANG BLK1/8INDEL
Supplier: IDEX HEALTH & SCIENCE LLC
Nut^ Flanged Fitting nut, Square, for 1/8in OD tubing, Color: Black, Material: Versatile Delrin, resists cross threading or loosening during use delivering excellent dimensional stability, It includes a 316 stainless steel washer. Compatible with a 1/4-28 Flat-Bottom port geometry
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NUT MALE FLANG BLK1/8INDEL 10803-774 P-501 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty