Would you like to log in again?
GRD COL UDC 2UM 120A KIT 2X10MM
Supplier: MicroSolv Technology Corp
Cogent Guard Column kit^ Cogent Guard Column Kit with UDC-Cholesterol 2.o, Pore Size: 120A Particle Size: 2.2um. Includes one Universal Holder and 5 each Hichrom Hardware Internal Diameter 2.0mm x 10mm Length Guard Columns.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GRD COL UDC 2UM 120A KIT 2X10MM 10803-798 69269-HG2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty