Would you like to log in again?
TUBING PEEK .010 ID 50' ROLL N
Supplier: MicroSolv Technology Corp
Tubing^ HPLC Tubing PEEK 0.010 inch ID x 1/16th inch OD. Natural 5000 psi max pressure. 50' Roll. Color Coded: Natural.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUBING PEEK .010 ID 50' ROLL N 10803-920 49350-10-N Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty