Would you like to log in again?
Screw Caps, MicroSolv™
Supplier: MicroSolv
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9504S-30C-W-K 9504S-35C-BLK 9504S-30C-W 9504S-80C-BK
10804-132PK 42.55 USD InStock
10804-132 10804-134 10804-136 10804-128
Screw Caps, MicroSolv™
Closures Chromatography Vial Closures
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty