Would you like to log in again?
SEPTA GREY SILICONE FOR HS 20MM CRIMP
Supplier: MicroSolv Technology Corp
Septa, Gray Silicone for crimp cap for Headspace. 3.175mm thick, 16mm diameter^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEPTA GREY SILICONE FOR HS 20MM CRIMP 10804-146 95025-24 Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty