Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Conductive Dividable Grid Containers, Quantum Storage Systems

Supplier: Quantum Storage Systems

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

DG93060CO DG91035CO DG92080CO DG92060CO DG92035CO DG91050CO DG93030CO DG93120CO DG93080CO
10804-291CS 588.42 USD InStock
10804-291 10804-293 10804-461 10804-295 10804-463 10804-297 10804-465 10804-467 10804-469
Conductive Dividable Grid Containers, Quantum Storage Systems
Boxes ESD-Safe Boxes
Protect your electronic components from typical electronmagnetic interference, electrostatic or induced fields with molded carbon-filled, copolymer polypropylene bins.

Conductive material provides a barrier which these fields cannot penetrate and prevents the build-up of electrostatic charge. Bins are permanently conductive and are unaffected by washing with normal cleaners.

Weigh capacity of 100 lbs. Divider available to customize your container into compartments at 11/8" Intervals.

Surface resistivity is less than 105 Ohms-per square. Black color.

Ordering information: Items must be ordered in increments of 12.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty