Would you like to log in again?
CAPS 10MM SCRW WHT SIL/PTFE SEPTA 1.25
Supplier: MicroSolv Technology Corp
Screw cap^ Screw cap with 1.25mm thick, silicone/PTFE septa fitted in a black cap, For use with 10-425, 12x32mm wide mouth vials, Basik brand
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS 10MM SCRW WHT SIL/PTFE SEPTA 1.25 10804-592 9503S-E-09C-K Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty