Would you like to log in again?
CAPS 9MM SCREW AQR 2MIL PTFE ONLY
Supplier: MicroSolv Technology Corp
Cap^ Cap, AQR, Autosampler, screw top, black with locked in fitted ultra pure 2mil PTFE only septa, For use with 9-425mm, 12x32mm AQ vials, AQ-Advanced Quality Brand
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS 9MM SCREW AQR 2MIL PTFE ONLY 10804-638 9509S-50C-B Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty