Would you like to log in again?
2ML CLR VIALS&BLK SIL/PTFE SEPTA CAPS
Supplier: MicroSolv Technology Corp
Vial^ Vial, 2ml, clear Borosilicate, wide mouth, glass 10-425, (12x32mm) Basik brand with black screw cap with fitted silicone/PTFE septa
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
2ML CLR VIALS&BLK SIL/PTFE SEPTA CAPS 10804-676 9503S-E-1CP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty