Would you like to log in again?
Puritan® Tapered Mini Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
823-WC 25-8265WC 25-8265WC 826-WC 826-WC
82004-051CS 874.71 USD InStock
82004-051 CA89133-754 CA10805-130 10805-130 89133-754
Puritan® Tapered Mini Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
This absorbent cotton tipped applicator is produced with high quality USP grade cotton, which is carded and coiled onsite to maintain consistency
It is bonded securely to a sturdy wood handle by an aqueous-based adhesive. The tapered mini cotton tip is ideal for hard-to-reach or small areas. Available nonsterile or sterile.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty