Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pur-Wraps® Swab, Cotton Tip, Sterile or Nonsterile, Puritan Medical Products
Supplier: Puritan Medical Products
Pur-Wraps®
25-8061PC 803-PC 803-PC 25-8061PC 806-PC 25-8062PC 25-8062PC
10805-148CS 695.05 USD InStock
10805-148 CA10805-144 10805-144 CA10805-140 10806-050 10805-140 CA10806-050
Pur-Wraps® Swab, Cotton Tip, Sterile or Nonsterile, Puritan Medical Products
Swabs Swabs for Sampling
This absorbent cotton tipped applicator is produced with high-quality USP grade cotton, which is carded and coiled onsite to maintain consistency.
It is bonded securely to a semi-flexible polystyrene handle by an aqueous-based adhesive. Available either nonsterile or sterile.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty