Would you like to log in again?
Puritan® Polyester Tip Applicator, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
25-8062WD 25-8062WD 25-8061WD 25-8061WD
89133-768BX 25.36 USD InStock
CA89133-768 CA10805-160 89133-768 10805-160
Puritan® Polyester Tip Applicator, Wood Handle, Sterile, Puritan Medical Products
Swabs Swabs for Sampling
Regular, non-absorbent polyester tip is bonded to a sturdy wooden handle with an aqueous adhesive
It is ideal for specimen collection, patient care, and surface collection. Made in the USA.
Certifications: CE marked.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty