Would you like to log in again?
Scout® Portable Topload Balance with Round Pan and Touchscreen Display, OHAUS®
Supplier: Ohaus
Scout®
30253007 30253008 30253009
10805-362EA 771.5 USD InStock
10805-362 10805-282 10805-280
Scout® Portable Topload Balance with Round Pan and Touchscreen Display, OHAUS®
Balances and Scales
Ideal for laboratory and industrial applications, the OHAUS® Scout® features an easy to navigate color touchscreen display and a slim design which allows stackable storage. Geared for superior performance with fast stabilization time and high resolution weighing results, these portable balances set a new standard for a broad range of weighing applications. Features also include superior overload protection and expanded connectivity options.
  • Informative Color Touchscreen Display
  • Superior Overload Protection and Stackable Storage
Additional standard features include; transportation lock, stainless steel pan, menu and calibration lockout switch, slip-resistant and adjustable feet, leveling bubble, mechanical and software overload/underload protection, stability indicator, auto tare, low battery indicator, auto shut-off, user selectable printing options, and user selectable communication settings.
Applications include Weighing, Parts Counting, Percent Weighing, Check Weighing, Animal/Dynamic Weighing, Totalization, Density Determination, Display Hold, and Mole Weighing.
Certifications: IEC/EN 61010-1; CAN/CSA C22.2 No. 61010-1; UL Std. No. 61010-1, IEC/EN 61326-1 Class B; FCC Part 15 Class B; Industry Canada ICES-003 Class B.
Ordering information: Includes AC adapter (4 AA batteries not included).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty