Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Precision™ General Purpose Water Baths, Thermo Scientific
Supplier: Thermo Scientific
TSGP2S TSGP20 TSGP28 TSGP15D TSGP02 TSGP10 TSGP05
10805-302EA 1370.48 USD InStock
10805-302 10805-300 10805-322 10805-298 10805-296 10805-306 10805-304
Precision™ General Purpose Water Baths, Thermo Scientific
Baths Water Baths, Unstirred
Support a wide range of applications with Thermo Scientific™ Precision™ general purpose water baths, designed to maintain water temperature from ambient to 100°C
These rugged, high-performance baths range from 2L to 28L, including shallow models. Over-temperature safety circuitry is designed to prevent thermal runaway, while new auto-on and auto-off timers allow you to optimize operation schedules. Benefit from outstanding chemical and corrosion resistance with epoxy powder-coated exterior, easily clean the chamber with its seamless stainless steel interior.
Certifications: cULus Listed, CE compliant, US FDA Class I Medical Device.
Delivery information: Supplied with Clear-Polycarbonate Lid, Diffuser Tray, Power Cord, Yellow Duck.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty