Economincal small parts storage systems feature open hopper front bins with a labeling area to provide fast access to stored items.
- All-in-one unit is easy to clean and will not rust or corrode
- Heavy-duty, high-grade shelving offers a 400 lbs. weight capacity per shelf
- 36" wide shelving has a smooth powder coat finish
Accessories information: Keep items separated in bin using dividers, provides additional storage options.
Ordering information: Bins are available in six colors.