Would you like to log in again?
12IN X 36IN X 39IN 24 BLUE 8IN BINS
Supplier: QUANTUM STORAGE SYSTEMS
Shelf System^ Economy 8 Inch Plastic Shelf bin System, Steel Shelving with plastic bins, 4 shelves, Number of Bins: 24, Heavy-Duty, Load: 400lbs capacity per shelf with smooth gray powder coat paint finish, Outer Dimater: 12x36x39 Inch, Bin size (LxWxH): 11-5/8 x 4-3/8 x 8 Inch, Color: Blue
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
12IN X 36IN X 39IN 24 BLUE 8IN BINS 10805-662 1239-SB801BL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty