Would you like to log in again?
COGENT HPLC COL AXIS CN 150 X 4.6 3UM
Supplier: MicroSolv Technology Corp
Column^ Column HPLC, Cogent Axis* Spherical Particles Length: 150 mm, Internal Diameter: 4.6 mm, Particle Size: 3 um, Chemistry: Axis Cyano, Pore Size: 100 A
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COGENT HPLC COL AXIS CN 150 X 4.6 3UM 10805-934 59034-15P-3 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty