Would you like to log in again?
COGENT F5 HPLC COLUMN 50 X 2.1MM
Supplier: MicroSolv Technology Corp
Column^ Column HPLC, Cogent F8 Spherical Particles Length: 50 mm, Internal Diameter: 2.1 mm, Particle Size: 5 um, Chemistry: F5 PentaFluroPhenyl, Pore Size: 60 A
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COGENT F5 HPLC COLUMN 50 X 2.1MM 10805-938 60022-05P-2 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty