Would you like to log in again?
SEPTA SIL/PTFE .4"THICK FOR 9M
Supplier: MicroSolv Technology Corp
Septa^ Septa only for autosampler vial cap. White silicone/ Natural PTFE for 9mm, Basik brand, ribbed screw cap, Cut to 0.40in thick and 0.346in diameter. Basik brand
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SEPTA SIL/PTFE .4"THICK FOR 9M 10805-994 96009-10-3010 Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty