Would you like to log in again?
Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
806-WCL 806-WCL 806-WC 806-WC
10806-001CS 294.53 USD InStock
CA10806-001 10806-001 10806-000 CA10806-000L
Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Swabs Swabs for Sampling
These Puritan® swabs are ideal for use in bacteriology labs for inoculating broth and plated media, and obtaining culture specimens.
Swabs with adhesive incorporate a nonsterile cotton tip secured by adhesive to the wooden handle. Swabs are also available without adhesive, for use where contamination of critical surfaces is a concern. Swabs are autoclavable.
They are also useful for cleaning delicate laboratory instruments.
Ordering information: Packaged in bags of 100 swabs, 10 bags to a pack, 10 packs to a case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty