Would you like to log in again?
PurSwab® Rectangular Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
PurSwab®
1805PFB-RECT 1805-PF-RECT 1805-PF-RECT 1805-PFBRECT
10806-032CS 856.67 USD InStock
CA10806-023 10806-032 CA10806-032 10806-023
PurSwab® Rectangular Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
Lint-free, nonabrasive, shred-resistant swabs are ideal for precision cleaning
Made in the USA.
Flat, slightly squared 100ppi polyurethane foam tip is securely bonded to the polypropylene shaft.
Certifications: CE marked.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty