Would you like to log in again?
VWR® Rochester Ocshner Kocher Rat Tooth Forceps
Supplier: VWR
VWR®
11647-117 11647-116 11647-120 11647-114 11647-118 11647-121 11647-119 11647-115
10806-102EA 39.43 USD InStock
10806-102 10806-104 10806-158 10806-098 10806-100 10806-160 10806-162 10806-106
VWR® Rochester Ocshner Kocher Rat Tooth Forceps
Forceps
VWR® Rochester Ocshner Kocher Rat Tooth Forceps are constructed of surgical grade stainless steel
  • Passivated
  • Rockwell Hardness (HRC): 40–45
  • Box-lock joint
  • Surgical grade
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
These instruments are available in curved or straight formats, in a variety of lengths. All forceps feature serrated jaws with rat tooth tips in a 1 x 2 teeth configuration. Forceps are fully autoclavable, and can be sterilized by EtO/gamma radiation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty