Would you like to log in again?
VWR® Premium Backhaus Towel Clamp
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11647-108
10806-124EA 73.51 USD InStock
10806-124
VWR® Premium Backhaus Towel Clamp
Clamps General Purpose Clamps
The VWR® Premium Backhaus Towel Clamp is constructed of German steel.
  • Passivated
  • Box-lock joint
  • Fully autoclavable
  • Surgical grade
  • Rockwell hardness (HRC): 40–45
This reusable clamp can be sterilized by EtO/gamma radiation.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty