Would you like to log in again?
PurSwab® Chemical Resistant Foam-Over-Cotton Tipped Applicator, Puritan Medical Products
Supplier: Puritan Medical Products
PurSwab®
1806-PCFCR 1806-WCF-CR 1806-WCFCR
10806-144CS 912.99 USD InStock
CA10806-144 CA10806-150 10806-144
PurSwab® Chemical Resistant Foam-Over-Cotton Tipped Applicator, Puritan Medical Products
Swabs Swabs for Sampling
This specialty foam tipped applicator is produced with chemical resistant foam.
Foam over cotton provides extra absorbency. It is slightly textured for light abrasion and will not swell or break apart with most solvents. For use in critical environments as a general cleaning swab, removing contamination and adhesives, and applying adhesives or other solutions.
Certifications: CE marked.
Ordering information: Packaged as 20 bags of 50 swabs per bag.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty