Would you like to log in again?
VWR® Rankin Kelly Hemostat Dissecting Forceps
Supplier: VWR
11647-127 11647-126
10806-168EA 39.8 USD InStock
10806-168 10806-166
VWR® Rankin Kelly Hemostat Dissecting Forceps
Forceps
VWR® Rankin Kelly Hemostat Dissecting Forceps are constructed of surgical-grade stainless steel.
  • Passivated
  • Box-lock joint
  • Fully autoclavable
  • Half-serrated jaws
  • Rockwell hardness (HRC): 40–45
These forceps can be sterilized by EtO/gamma radiation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty