Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
VWR® Pean Hemostatic Forceps
Supplier: VWR
VWR®
11647-150 11647-142 11647-139 11647-144 11647-140 11647-138 11647-146 11647-147 11647-151 11647-136 11647-149 11647-135 11647-145 11647-143 11647-148 11647-141 11647-134 11647-137
10806-134EA 120.83 USD InStock
10806-134 10806-200 10806-202 10806-186 10806-130 10806-196 10806-210 10806-188 10806-198 10806-208 10806-128 10806-204 10806-206 10806-182 10806-192 10806-184 10806-194 10806-190
VWR® Pean Hemostatic Forceps
Forceps
VWR® Pean Hemostatic Forceps are constructed of surgical grade stainless steel
  • Passivated
  • Box-lock joint
  • Surgical grade
  • Rockwell Hardness (HRC): 40–45
  • Fully autoclavable
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
These instruments are compatible with a variety of sterilization methods, and are available in an assortment of lengths and jaw types.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty