Would you like to log in again?
VWR® Lempert Bone Rongeur
Supplier: VWR
VWR®
11647-234 11647-233
10806-246EA 332.33 USD InStock
10806-246 10806-248
VWR® Lempert Bone Rongeur
Bone Cutters and Rongeurs
The VWR® Lempert Bone Rongeur is made from ASTM (899-94) Series 400 German stainless steel.
  • Passivated
  • Surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in a straight or curved format, this insturment is designed for cutting cartilage and bone. A strong, removable bar-spring makes these shears self-opening.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty