Would you like to log in again?
VWR® Micro Cleveland Bone Rongeur
Supplier: VWR
VWR®
11647-240 11647-239
10806-258EA 332.22 USD InStock
10806-258 10806-260
VWR® Micro Cleveland Bone Rongeur
Bone Cutters and Rongeurs
The VWR® Micro Cleveland Bone Rongeur is made from ASTM (899-94) Series 400 German stainless steel.
  • Passivated
  • Surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
The instrument is designed for cutting cartilage and bone, and feaures a strong, removable bar-spring that makes these shears self-opening.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty