Would you like to log in again?
VWR® Spear Shaped Dissection Needle
Supplier: Avantor
VWR®
11647-168 11647-169
10806-322EA 30.61 USD InStock
10806-322 10806-324
VWR® Spear Shaped Dissection Needle
Dissection Needles
VWR® Spear Shaped Dissection Needles are made from ASTM (899-94) Series 400 Stainless Steel.
  • Passivated
  • Hexagonal
  • Stainless steel handle
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Needles are available with either a single-cutting edge, or a double-cutting edge. Each format features a stainless steel handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty