Would you like to log in again?
VWR® Dissection Needle with Adjustable Chuck
Supplier: Avantor
VWR®
11647-172
10806-330EA 13.13 USD InStock
10806-330
VWR® Dissection Needle with Adjustable Chuck
Dissection Needles
The VWR® Dissection Needle with Adjustable Chuck is made from ASTM (899-94) Series 400 Stainless Steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Dissection needles feature an adjustable chuck to tighten or loosen steel needles, and comes with one straight needle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty