Would you like to log in again?
VWR® Graefe Micro Hook
Supplier: Avantor
VWR®
11647-177 11647-178
10806-340EA 51.64 USD InStock
10806-340 10806-342
VWR® Graefe Micro Hook
Dissection Needles
The VWR® Graefe Micro Hook is constructed from German steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Devices are available in sharp or blunt formats.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty