Would you like to log in again?
VWR® One-Piece Zahn Cup-Style Viscometers
Supplier: VWR
VWR®
11647-201 11647-200 11647-203 11647-204 11647-202
10806-244EA 292.39 USD InStock
10806-244 10806-362 10806-240 10806-364 10806-242
VWR® One-Piece Zahn Cup-Style Viscometers
Viscometers Flow Cup Viscometers
VWR®One-Piece Zahn Cup-Style Viscometers are constructed of corrosion-resistant, ASTM (899-94) Series 400 Stainless Steel.
  • Passivated
  • Compatible with Gamma/ETO/autoclavable sterilization methods
  • Corrosion-resistant
Viscometers provide measurement of liquids ranging from very thin to very heavy. All devices are supplied with a Calibration Certificate.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty