Would you like to log in again?
VWR® Vial Decapper Pliers
Supplier: VWR
VWR®
11647-210 11647-205 11647-213 11647-207 11647-206 11647-214 11647-211 11647-208 11647-212 11647-209
10806-366EA 137.33 USD InStock
10806-366 10806-376 10806-368 10806-378 10806-384 10806-372 10806-374 10806-380 10806-382 10806-370
VWR® Vial Decapper Pliers
Crimpers, Cappers and Decappers
VWR® Vial Decapper Pliers are designed to easily remove aluminum crimp tops from vials and bottles.
  • Passivated
  • Surgical-grade stainless steel construction
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in single- or double-action formats, all pliers are smooth and simple to operate. Durable, high-quality construction ensures consistent, dependable performance.
Overall length: 159 mm (6.25")
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty