Would you like to log in again?
VWR® Pure Nickel Dish
Supplier: Avantor
VWR®
11647-217
10806-390EA 102.98 USD InStock
10806-390
VWR® Pure Nickel Dish
Trays/Dishes/Pots Laboratory Trays/Dishes
The VWR® Pure Nickel Dish is made from a one-piece ASTM standard (A-494) Pure Nickel sheet, spun to shape
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Dishes feature straight sides, with a lip for pouring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty