Would you like to log in again?
R-300H IP WB 1L 24/40 V PG 110
Supplier: Buchi
Rotavapor^ Rotavapor R-300H, Lift System: Manual, Heating Bath: B-301 (20 - 95 deg, 1L), Standard Joint: SJ 24/40, Glass Assembly: Vertical (V), Interface: I-300 Pro, valve, P+G safety coating, Dimension: 400 x 615 x 320 mm, Frequency: 50/60 Hz, Rotation speed: 10 - 280 rpm, Voltage: 110V
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
R-300H IP WB 1L 24/40 V PG 110 10806-958 11R300112V132 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty