Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Rotavapor® R-300M Rotary Evaporators with Controller, BUCHI
Supplier: Büchi
Rotavapor®
11R300152V122 11R300152C152 11R300152V142 11R300152V152 11R300152C132 11R300152C122 11R300152V132 11R300152C142
10807-186EA 13961.92 USD InStock
10807-186 10807-250 10807-184 10807-246 10807-244 10807-188 10807-190 10807-248
Rotavapor® R-300M Rotary Evaporators with Controller, BUCHI
Evaporators Rotary Evaporators
The R-300 meets the highest expectations in convenience and versatility.
  • Intuitive user interaction
  • Precise operation control
  • 5 L flask capacity
  • Ergonomic handle
  • Combi-clip
  • Additional glass assemblies and configurations
The R-300 Rotavapor® has an ergonomic handle for easy positioning of the evaporation flask in to and out of the water bath. The I-300 interface is the central control unit for all process parameters of the Rotavapor R-300. This includes rotation speed; lift position; heating, cooling, and vapor temperature and pressure control. Push notifications on mobile devices allows connected users to view the status of multiple R-300s. The patented multipurpose Combi-clip with snap lock-mechanism provides for quick flask connection and detachment with one hand.
The I-300 interface provides a central location to regulate each evaporation parameter.
Certifications: CE and CSA compliant.
Accessories information: I-300 standard or I-300 Pro controller interface included.
Ordering information: All accessories and connections are included for out of the box usage. Optional 1L bath, safety shield, 1L evaporation flasks, different joint size glassware, valves for in-house cooling and vacuum are available as accessories (sold separately). Additional glass assemblies and configurations are available. Please contact customer service for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty