Would you like to log in again?
VWR® Sponge Holding Forceps
Supplier: VWR
11647-156 11647-154 11647-158 11647-157 11647-155
10807-450EA 60.41 USD InStock
10807-450 10807-444 10807-452 10807-448 10807-446
VWR® Sponge Holding Forceps
Forceps
VWR® Sponge Holding Forceps feature oval, fenestrated, serrated tips.
  • Oval, fenestrated, serrated tips
  • Suited to holding gauze squares
  • Ratcheted locking mechanism
These forceps are commonly used to hold gauze squares that will absorb excess fluids and blood from hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty