Would you like to log in again?
V-300 Vacuum Pump, Standalone, BUCHI
Supplier: Büchi
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11V300100 11V300110 11V300000 11V300000 11V300110 11V300100
10808-692EA 5258.39 USD InStock
10808-692 CA10808-692 10023-974 10808-700 CA10023-974 CA10808-700
V-300 Vacuum Pump, Standalone, BUCHI
Pumps Vacuum Pumps
The chemically resistant PTFE diaphragm vacuum pump impresses with its quiet and economical operation.
  • Quiet operation
  • Oil-free PTFE diaphragm pump
  • Reliable
  • Visible operating membranes
The V-300 vacuum pump works very quietly at 32 dB, with PTFE membranes visible to monitor presence of any condensed solvent inside the pump.
Certifications: CE, CSA and CUS compliant.
Ordering information: All accessories and connections are included for out of the box usage. Waste level sensors are sold separately; please contact your VWR representative for more information. Additional accessories are required for adaptation with older BUCHI systems. Please contact your VWR representative for further assistance.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: BUCHI provides convenient PM Mail - in services. After ordering, you will receive a protective box to ship your equipment to BUCHI Corporation along with a packaging instruction. Upon receipt, your equipment will be inspected for any defects and the performance of your instrument will be compared to manufacturer standards. After the PM, a service report and your equipment will be carefully repacked then returned to you with standard ground shipping.
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty