Would you like to log in again?
3M™ GoggleGear™ 500 Series, 3M™ Company
Supplier: 3M
3M™
7100079187
10808-716CS 267.46 USD InStock
10808-716
3M™ GoggleGear™ 500 Series, 3M™ Company
Goggles
GoggleGear™ 500 Series offers a low-profile design with an adjustable strap and indirect ventilation.
  • Long lasting Scotchgard™ anti-fog coating helps to increase eyewear performance in wet and steamy environments
  • Workers enjoy reliable, clearer sight longer, as the coating and its active ingredient is bonded to the lens
  • Coating retains its effectiveness for at least 25 washings with water
  • The coating withstands disinfection with diluted bleach soaking or alcohol wipes without losing anti-fog performance
Scotchgard™ anti-fog technology brings higher anti-fog performance to safety eyewear, helping workers see more clearly to complete on-the-job tasks. The anti-fogging properties are based on internal testing per EN168 test method, when compared to traditional anti-fog coatings. When microscopic water droplets land on lenses without any type of coating, they bead and form condensation and fog, obscuring vision. With cotchgard™ anti-fog technology, the anti-fog coating produces a reduced-contact angle, flattening water beads into a thin, transparent film that allows light to pass through. The coating resists fogging longer than traditional anti-fog coatings, even after multiple washes.
3M™ Scotchgard™ anti-fog coating, with its active ingredient bonded to the lens, helps the eyewear last longer. Eyewear also comes with polycarbonate lenses that absorb 99.9% of UV rays.
Certifications: Meets ANSI Z87.1 and CSA Z94.3.
Accessories information: GoggleGear™ 500-Series Prescription Insert is helpful for any users requiring corrective vision to go along with eye protection.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty