Would you like to log in again?
BIOMEK 4000 ACCU-FRAME ADAPTER KIT
Supplier: BECKMAN COULTER
Biomek* 4000 AccuFrame kit^ Biomek* 4000 AccuFrame Kit, Used on the main deck and side module positions. Off-Deck Tool Rack, Off-Deck Gripper Rack, and Liquid Waste Stations must be framed manually.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIOMEK 4000 ACCU-FRAME ADAPTER KIT 10808-910 B22868 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty