Would you like to log in again?
ACCY LEFT SIDE MODULE
Supplier: BECKMAN COULTER
Module Accessory^ Module Accessory, Left Side, for Biomek* 3000 worksurface, side module cover is removable for easy cleaning, and a side module spill tray. Side module holds two labware or pipette tip rack holders, two tool racks, or one test tube rack holder.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ACCY LEFT SIDE MODULE 10808-914 987264 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty