Would you like to log in again?
BIOMEK 4000 LIQUID WASTE DISPOSAL KIT
Supplier: BECKMAN COULTER
Biomek* 4000 Liquid Waste Disposal kit^ Biomek* 4000 Liquid Waste Disposal Kit, for liquid waste disposal fed either off or on deck, Can be used in off deck location to save deck space, Includes liquid drain, tubing, and waste bottle
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BIOMEK 4000 LIQUID WASTE DISPOSAL KIT 10808-922 B21398 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty