Would you like to log in again?
P20 TOOL 1-TIP B3K
Supplier: BECKMAN COULTER
P20 Tool^ P20 Tool, 1-Tip, Biomek* 3000, MP20 Eight-Tip Pipette Tool, Capacity: 1 to 20 uL for each tip, Includes true 9 mm center-to-center spacing and can be used with SBS/ANSI standard labware.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
P20 TOOL 1-TIP B3K 10808-930 987367 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty