Would you like to log in again?
PolarSafe™ Aluminum Cooling Blocks, Argos Technologies
Supplier: Argos Technologies
PolarSafe™
63615-03 63615-01 63615-05 63615-04 63615-02
10808-958EA 220.7 USD InStock
10808-958 10808-960 10808-952 10808-954 10808-956
PolarSafe™ Aluminum Cooling Blocks, Argos Technologies
Boxes Cooling/Freezing Boxes and Bags
PolarSafe™ Cooling Blocks are perfect for the utmost in temperature control for precious laboratory samples
Uniform well construction provides inserted tubes with a snug fit and a highly efficient degree of thermal conductivity. Simply place the PolarSafe™ cooling blocks directly onto the desired temperature source and they will quickly adapt to the new temperature.
Whether cooling, freezing, or heating (thawing), these blocks will provide welcome temperature control. Ideal for use on ice, dry ice, in water baths, and in liquid nitrogen.
The family of 5 racks will help keep biological samples upright, organized, and at uniform temperatures for extended periods of time. There are options to hold 1.5/2.0mL MCT’s and cryogenic vials, 0.5mL tubes, as well as 96- and 384-well plates.
These electric blue aluminum blocks are anodized to stand up to cold temperatures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty