Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Thermoshaker for Microplates, Grant Instruments
Supplier: Grant
PHMP-100USA
10809-088EA 4456.79 USD InStock
10809-088
Thermoshaker for Microplates, Grant Instruments
Shakers and Mixers Shaking Incubators
The PHMP-100 is a thermoshaker with excellent temperature uniformity across the platform/microplate (due to the bi-directional heating system) combined with variable speed and variable temperature produces the ideal thermoshaker for microplate incubations up to 100°C.
  • Lid lock and safety interlock causing the rotor to stop when lid is opened.
  • Multi-spin enables consecutive spin and mix phases of multiple tubes
  • Choice of interchangeable rotors for different microtube sizes/combinations and for the 0.2ml strip
Average heat up speed is 12 minutes from 25°C to 37°C, 35 minutes from 25°C to 60°C, and 60 minutes from 25°C to 100°C.
Continuous or timed operation, with alarm buzzer and automatic switch-off facility.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty